Volumetric flasks, class B, red markings
Supplier: VITLAB
67695
VITL67695EA 63.2 SGD
VITL67695
Volumetric flasks, class B, red markings
Flasks Volumetric Flasks
PMP, translucent, with PP NS-stopper.
  • Tolerances are approximately 20% better than class B according to DIN EN ISO 1042
  • Individually ring marked
  • With NS stoppers
Certifications: DIN EN ISO 1042
Order Now
SPECIFICATIONS
Environmentally Preferable