Hydrometer cylinders
Supplier: VITLAB
760941
VITL760941EA 83.8 SGD
VITL760941
Hydrometer cylinders
Cylinders Hydrometer Cylinders
Cylinder, made of PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Robust and stable
With spout and overflow vessel. This ensures that the hydrometer can be read with a completely filled cylinder.