Spatulas, PA
Supplier: VITLAB
80596
VITL80596EA 31.7 SGD
VITL80596
Spatulas, PA
Spatulas
Double-ended spatulas made of glass-fibre reinforced PA.
  • With stable, easy to hold handle in the middle
  • Two spatula endings or one spoon and one spatula ending
Order Now
SPECIFICATIONS
Environmentally Preferable