Wash bottles, narrow or wide neck
Supplier: VITLAB
95188 94688 94188
VITL95188EA 252 SGD
VITL95188 VITL94688 VITL94188
Wash bottles, narrow or wide neck
Bottles Wash Bottles
Made of LDPE, translucent, screw cap and spray tube from PP.
  • Good chemical resistance
  • Good temperature resistance
Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.
Order Now
SPECIFICATIONS
Environmentally Preferable