VWR® SilkSkin®, Examination Gloves, Nitrile
Supplier: VWR Collection
VWRA112-3384EA 40.9 SGD
VWRA112-3384 VWRA112-3386 VWRA112-3385
VWR® SilkSkin®, Examination Gloves, Nitrile
Gloves
VWR® SilkSkin® examination gloves are ideal for users with the most sensitive skin, and allergy sufferers, as the anti-aging effects regenerate, moisturize, nourish, and smooth the skin.
  • UV, anti-microbial, and wound healing properties
  • Smooth palms with textured fingers
  • Beaded cuffs
  • Single-use design
  • Non-sterile
VWR® SilkSkin® examination gloves provide unique benefits to wearers suffering from dry or sensitive skin types. Like silk on the skin, SilkSkin® examination gloves feature natural moisturizing factors that hydrate and soften the hands.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Disposable / reusable Disposable
Latex / latex-free Latex-free
Powdered / powder-free Powder-free
Material Nitrile
Finish Textured fingers
Cuff style Beaded
Finger thickness 0,08 mm
Palm thickness 0,07 mm
Anti-static +