VWR®, Tissue Homogeniser
Supplier: VWR Collection
VWRA432-0430EA 7520 SGD
VWRA432-0430
VWR®, Tissue Homogeniser
Homogenisers Homogenisers Bench
This homogeniser has been developed and designed after extensive R and D and perfected with stringent quality checks. It is equiped with components such as the powerful maintenance-free BLDC motor which offers exceptional performance together with high sample output and better shearing force.
  • Ideal homogeniser for extraction from samples like bacterial/yeast cells, plant tissues and animal tissues for molecular analysis
  • Accommodates wide range of screw cap homogenising tubes sizes 24×2 ml and 12×7 ml with adaptors
  • Designed to generate less heat and have
More Product Information