VWR®, Economical Junior Stirrer
Supplier: VWR Collection
VWRA441-0509EA 977 SGD
VWRA441-0509
VWR®, Economical Junior Stirrer
Stirrers Overhead Stirrers
The Junior series of stirrers combines a compact form factor with exceptional performance and enhanced convenience. Various built-in safety features ensure that no matter what, your safety is never compromised. The instrument has a brushless DC motor delivering ultra-quiet performance and maintenance which hits the electronic speed from 50 - 2200 rpm
  • Microprocessor controlled for consistent accuracy with viscous and non-viscous liquids
  • Stirs viscosity up to 1000 mPas or 10 L volume capacity (H2O)
  • Adjustable speed ranging from 50 Up to 2200 RPM
  • Safety cut
More Product Information