VWR®, Prime Stirres
Supplier: VWR Collection
VWRA441-0508EA 696 SGD
VWRA441-0508 VWRA441-0507 VWRA441-0510 VWRA441-0511
VWR®, Prime Stirres
Stirrers Overhead Stirrers
Stirrers which are ideal for laboratory stirring tasks with perfect blend of exceptional performance from our prime range of stirrers, models have a compact design and constant torque in four different version with 8, 15, 20 and 40 Ncm and adjustable speed ranging from 50 rpm to 2000 rpm.
  • Microprocessor controlled for maintaining accuracy even with viscous liquid
  • Viscosity up to 8000 mPas and 20 L volume
  • Counter reaction to speed change
  • Push through shaft for viscous liquid or smaller vessels
  • Long timer setting from 1 min to 99 hours 59 mins and continuous
More Product Information
Order Now
SPECIFICATIONS
Model Overhead stirrer 20 Ncm Overhead stirrer 40 Ncm Overhead stirrer 8 Ncm Overhead stirrer 15 Ncm
Max. stirring capacity H₂O 25 L 40 L 10 L 20 L
Environmentally Preferable
Max. viscosity 10000 mPas 30000 mPas 4000 mPas 8000 mPas
Speed range 2000 rpm 1000 rpm 2000 rpm 1000 rpm
Max. torque 20 Ncm 40 Ncm 8 Ncm 15 Ncm