VWR®, Economical Vortex Mixer
Supplier: VWR Collection
VWRA444-1146EA 207 SGD
VWRA444-1146
VWR®, Economical Vortex Mixer
Shakers Vortex Shakers
Laboratory vortex mixer for quick and convenient mixing of small sample volumes. Compact mixer with a wide range of intutive features.
  • Tiny vortex mixer with a small footprint
  • Delivers up to 3500 min⁻¹ for thorough mixing and quick vortexing
  • Smart and robust design - press activation with suspension
  • Robust construction with die cast aluminum base and rubber vaccum pads for stable and silent operation
  • Built-in counterbalance for stable operation
Certifications: IP Protection Class according to DIN EN 60259: IP 43.
Delivery information: Supplied with universal attachment.