VWR®, Digital Plate Tube Rollers
Supplier: VWR Collection
VWRA444-1178EA 789 SGD
VWRA444-1178
VWR®, Digital Plate Tube Rollers
Shakers Rotating/Rocking Shakers
With quality internal components including a powerful, maintenance-free brushless DC motor and features such as power intermittent mode and user defined programming functions. This plate rotator provides exeptional performance every single time with 100% reliability.
  • Microprocessor controlled plate rotator with variable speed and time setting, features last run memory function
  • Programmable – user can set and save up to 9 user-defined programs (protocols)
  • Pulse mode function for alternating clockwise and anticlockwise motion with 2 second stops for enhanced,
More Product Information
Order Now
SPECIFICATIONS
Capacity 16×50 ml tubes
Environmentally Preferable
Speed 10 - 80 min⁻¹
Angle 0 - 90°
W×D×H 540×274×186 mm
Timer 1 - 99 min and infinite mode