VWR® Motorless Ultra Slim Magnetic Stirrer, Single Position
Supplier: VWR Collection
VWRA444-1189EA 296 SGD
VWRA444-1189
VWR® Motorless Ultra Slim Magnetic Stirrer, Single Position
Stirrers Magnetic Stirrers
Developed to offer reliable performance and years of durability with a motorless design. The design offers exceptional performance without the hassle of maintenance.
  • No moving parts-truly maintenance free
  • Low aspect ratio (just 13/14 mm thick)
  • Microprocessor controlled and last run memory function
  • Pulse mode - reverses the direction every 30 seconds for proper mixing
  • Software driven gradual acceleration
  • Universal power supply - small wall mount adaptor
  • IP 65 complaint design (Water spill resistant)
  • Corrosion resistant SS top surface for better chemical
More Product Information