VWR® Motorless Ultra Slim Magnetic Stirrer, 4 Position
Supplier: VWR Collection
VWRA444-1191EA 696 SGD
VWRA444-1191
VWR® Motorless Ultra Slim Magnetic Stirrer, 4 Position
Stirrers Magnetic Stirrers
This range of magnetic stirrer provides reliable performance and maintenance-free long life. With an aspect ratio of just 13 mm thick and weighing just 2,5 kg. This stirrer is perfect representation of immense power and a compact package.
  • No moving parts-truly maintenance free
  • Microprocessor controlled and last run memory function
  • Universal power supply- small wall mount adaptor
  • Corrosion resistant SS top surface for better chemical resistance
  • Synchronised speed setting (multiposition)
With convince built right into the design and user interface ofMore Product Information