VWR® Analog Magnetic Stirrer
Supplier: VWR Collection
VWRA444-1195EA 188 SGD
VWRA444-1195
VWR® Analog Magnetic Stirrer
Stirrers Magnetic Stirrers
Economical product range without compromising on flexibility or durability, with its stainless steel top surface, the device is extremely resistant to corrosion while its long lasting brushless DC motor runs at top speed without needing much of a maintenance.
  • Variable speed ranging from 400 to 2200 min⁻¹
  • Closed design for protection against accidental spillage and easy cleaning
  • Easy to operate-plug and play
  • Non slip strength rubber feet
  • Small footprint for saving valuable bench space
  • Suitable for use in hoods and cold rooms
  • IP 42 protection class