VWR® Stirrers 2/4/6 L Capacity
Supplier: VWR Collection
VWRA444-1212EA 1170 SGD
VWRA444-1212 VWRA444-1213 VWRA444-1214
VWR® Stirrers 2/4/6 L Capacity
Stirrers Magnetic Hotplate Stirrers, Multi-position
Digital hot plate stirrers with 5, 10, and 15 stirring positions are engineered to offer unparalleled efficiency. Both the software and hardware of these devices are tailored to maximize productivity and safety with their exceptional performance. They come equipped with a programmable pulse and a heating efficiency mode for an intuitive user experience.
  • High performance multi position stirrer with 5, 10 and 15 stirring positions with optimum use of laboratory space
  • Variable speed ranging from 300 to 1500 min⁻¹ in steps of 10 min⁻¹
  • Timer setting from 1 to 99
More Product Information
Order Now
SPECIFICATIONS
Model VWRA444-1212 VWRA444-1213 VWRA444-1214
Environmentally Preferable
Plate dimensions 565×136×75 mm 565×196×75 mm 565×282×75 mm
No. of stirring positions 5 10 15
Speed range 300 - 1500 min⁻¹ 300 - 1500 min⁻¹ 300 - 1500 min⁻¹
Timer 1 - 999 min. and infinite (run time) 1 - 999 min. and infinite (run time) 1 - 999 min. and infinite (run time)
Temperature range Ambient…120 °C Ambient…120 °C Ambient…120 °C