VWR®, Microprocessor and Brushless Motor High Capacity Magnetic Stirrer 50 L
Supplier: VWR Collection
VWRA444-1216EA 1880 SGD
VWRA444-1216
VWR®, Microprocessor and Brushless Motor High Capacity Magnetic Stirrer 50 L
Stirrers Magnetic Stirrers
The 50 L magnetic stirrer with brushless motor is equipped with top range features which delivers exceptional stirring performance even under max capacity.
  • Adjustable speed ranging from 100 to 1000 min⁻¹ 50 L with increment or decrement of ±10 min⁻¹
  • Long timer setting from 1 to 99 hr and 59 min and infinite mode
  • Choice of acceleration rates (A1-A9 with A9 being the fastest rate and A1 slowest)
  • Programmable pulse mode with direction change feature
  • Alternating clockwise and anticlockwise motion with 3 sec stop for better and thorough mixing
  • Stainless steel
More Product Information