VWR®, Microprocessor and Brushless Motor High Capacity Magnetic Stirrer 200 L
Supplier: VWR Collection
VWRA444-1219EA 2300 SGD
VWRA444-1219
VWR®, Microprocessor and Brushless Motor High Capacity Magnetic Stirrer 200 L
Stirrers Magnetic Stirrers
The high quality components such as the brushless DC motor and microprocessor control ensures powerful and efficient performance to deliver exceptional, while using large heating solutions that need to be magnetically stirred these high capacity magnetic stirrers are ideal tools for the tasks.
  • Adjustable speed ranging from 100 to 1000 min⁻¹ with increment or decrement of ±10 min⁻¹
  • Long timer setting from 1 to 99 hr and 59 min and infinite mode
  • Ramp up speed feature: choice of acceleration rates from A1 to A9 (with A9 being the fastest rate and A1 the
More Product Information