VWR®, Personal microplate shaker, single position
Supplier: VWR Collection
VWRA444-1221EA 395 SGD
VWRA444-1221
VWR®, Personal microplate shaker, single position
Shakers Microplate Shakers
This device has been designed to offer high performance and exceptional ease-of-use in a compact package.
  • Microprocessor controlled for efficient performance even under high loads
  • Variable timer setting from 1 to 99 minutes and infinite mode
  • Compact footprint saves valuable space and can also be used inside cold rooms/incubators
  • Digital display
  • With in-built counter balance
The powerful brushless DC motor and variable parameter settings makes it an amazingly versatile device to suit various laboratory requirements.
Delivery information: SuppliedMore Product Information