VWR®, Digital 4 Plate Incubated Shaker
Supplier: VWR Collection
VWRA444-1223EA 1640 SGD
VWRA444-1223
VWR®, Digital 4 Plate Incubated Shaker
Shakers Microplate Shakers
This device has been deisgned for performance and durability, with the objective of delivering perfectionism. One of the key feature is the maintenance free brushless DC motor, which performs efficiently even under high loads for thorough mixing.
  • Temperature setting range from Ambient +3 to 70 °C
  • Adjustable speed ranging from 200 to 1200 min⁻¹
  • Microprocessor controlled Thermo Shaker with variable speed and time setting and also with last run memory function
  • Long timer setting up to 99 hours and 59 minutes
  • Critical parameters like speed, time as well as set
More Product Information