VWR®, Orbital Platform Shaker
Supplier: VWR Collection
VWRA444-1231EA 1610 SGD
VWRA444-1231
VWR®, Orbital Platform Shaker
Shakers Orbital Shakers
This platform shaker combines performance with user-friendliness for enhanced convenience. With a powerful brushless DC motor and top-of-the line components, it is fully equipped to deliver exceptional performance. Leading features like loop function and user programmable interface ensure the device is convenient and easy to use.
  • Microprocessor controlled for efficient performance even with heavy loads
  • Variable speed selection up to 300 min⁻¹
  • Optional RS232 interface for remote operation
  • Auto power on in case of power failure
Certifications: IPMore Product Information