VWR®, Analogue Magnetic Hotplate Stirrer
Supplier: VWR Collection
VWRA444-1282EA 320 SGD
VWRA444-1282
VWR®, Analogue Magnetic Hotplate Stirrer
Stirrers Magnetic Hotplate Stirrers
Tested extensively to ensure efficient performance and an extremely user-friendly interface, VWR® Analogue Hot Plate Stirrer has been heavily researched in its development phase. With the best features, it is the solution to all your hot stirring needs with a capacity of up to 10 litres. Additionally, due to its small footprint it saves valuable bench space and is suitable for use in hoods and cold rooms.
  • Powerful Servo DC motor deliver up to 2200 RPM operation and maintenance free long operation.
  • Variable speed ranging from 500 - 2200 RPM
  • Temperature
More Product Information