VWR® Ultra Low Temperature Freezers, Smart Frequency Series
Supplier: VWR Collection
VWRA471-1281EA 26710 SGD
VWRA471-1281 VWRA471-1283 VWRA471-1284
VWR® Ultra Low Temperature Freezers, Smart Frequency Series
Freezers Ultra Low Temperature Freezers
Applicable for products and samples which require strict storage conditions such as viruses, pathogens, red blood cells, white blood cells, skin, bones, bacteria, semen, biological products, electronics and special materials.
  • Smart full-size 10" touch screen with state-of-the-art user interface
  • HC and Variable Frequency Drive Refrigeration System for Additional Energy Saving Advanced innovative design delivers superior energy savings
  • Advanced innovative design delivers superior energy savings
  • The energy consumption is down to a single digit
  • Check the real-
More Product Information
Order Now
SPECIFICATIONS
Model 471-1281 471-1283 471-1284
Environmentally Preferable
Display LCD Touchscreen LCD Touchscreen LCD Touchscreen
Alarm High/Low Temperature
Hot Condenser
Power Failure
High/Low Voltage
Sensor Error
Low Battery
High Ambient Temperature
Door Ajar
High/Low Temperature
Hot Condenser
Power Failure
High/Low Voltage
Sensor Error
Low Battery
High Ambient Temperature
Door Ajar
High/Low Temperature
Hot Condenser
Power Failure
High/Low Voltage
Sensor Error
Low Battery
High Ambient Temperature
Door Ajar
Weight 385 kg 415 kg 485 kg