VWR®, Personal Micro-Centrifuge
Supplier: VWR Collection
VWRA521-2778EA 203 SGD
VWRA521-2778
VWR®, Personal Micro-Centrifuge
Centrifuges Microcentrifuges
Made from the highest quality of raw materials and components, this micro-centrifuge will fit seamlessly into your work life and deliver exceptional performance. Designed to offer optimum performance and years of durability, the unit has been especially tailored to provide enhanced airflow to prevent the rise of temperature inside the chamber.
  • Electric brake for stopping the device immediately
  • Magnetic design for tool-free rotor removal
  • Carbon dust insulation and safe drainage provision
  • Quick run function to save time
  • Speed accuracy of ±200 min⁻¹