VWR®, Digital disc rotators
Supplier: VWR Collection
VWRA521-2779EA 752 SGD
VWRA521-2779
VWR®, Digital disc rotators
Shakers Rotating/Rocking Shakers
The Digital disc rotator is the result of extensive research and user experience feedback. Consisting of a maintenance free brushless DC motor, the rotator offers powerful performance without any hassel. The features such as intermittent mode, user programmable setup and its pulse mode offer thorough mixing without much effort.
  • Microprocessor controlled disc rotator for intelligent performance with last run memory feature
  • Variable angle setting to adjust the rotator angle from 0 to 90°
  • User programmable mode to set 9 user-defined programs for greater
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Speed 10 - 80 min⁻¹
Tilt angle 0 - 90°
W×D×H 383×325×325 mm
Timer 1 - 99 min and infinite mode