VWR®, Magnetic Hotplate Stirrer, 320 ℃
Supplier: VWR Collection
VWRA521-2782EA 470 SGD
VWRA521-2782
VWR®, Magnetic Hotplate Stirrer, 320 ℃
Stirrers Magnetic Hotplate Stirrers, Multi-position
With class leading features that are designed to deal with demanding laboratory conditions and real life situation, this equipment is the answer to all of your hotplate stirring needs.
  • Max stirring capacity of 10 L
  • Heater temperature setting from ambient to 320 °C with safe temperature setting upto 345 °C
  • Safe temperature circuit for accurate temperature during operation
  • Variable speed setting from 200 to 2200 min⁻¹ in steps of 10 min⁻¹
  • Long timer range from 1 to 999 mins and infinite mode
It has been equipped with top of the line features whichMore Product Information