VWR®, Personal Microcentrifuge
Supplier: VWR Collection
VWRA521-2794EA 320 SGD
VWRA521-2794
VWR®, Personal Microcentrifuge
Centrifuges Microcentrifuges
Following in-depth user research, this personal micro-centrifuge with eight slots is designed to meet all user expectations. It offers dependable performance and high-quality parts, combined with a smartly designed frame and an intuitive user interface, all aimed at simplifying laboratory work.
  • Microprocessor controlled for efficient performance
  • Imbalance detection
  • Compact yet robust design saves valuable bench space
  • Highly durable thanks to robust construction
  • Can be used at 5 to 50 °C (80% relative humidity), in a fume hood or cold room
  • Variable speed
More Product Information