VWR® Mini 12, Mini Centrifuge
Supplier: VWR Collection
VWRA521-2795EA 1360 SGD
VWRA521-2795
VWR® Mini 12, Mini Centrifuge
Centrifuges Microcentrifuges
This micro-centrifuge is designed and developed to provide optimum performance along with exceptional ease of use. The use of brushless DC motor and intelligent design provide enhanced airflow resulting in an excellent user-experience.
  • Bright and clear LCD display
  • Small footprint saves valuable bench space
  • Intelligent design ensures smooth and silent operation even at maximum speed
  • Dual shell design and tested explosion proof construction
  • Safer low voltage device (24 V) with universal power supply
With features like lid lock safety, one touchMore Product Information