VWR® Combi Centrifuge
Supplier: VWR Collection
VWRA521-2797EA 1130 SGD
VWRA521-2797
VWR® Combi Centrifuge
Centrifuges Benchtop Centrifuges
Unit has a combi rotor that works effectively without generating much noise and heat.
  • Combi rotor accommodates 12×1,5/2 ml and 4×PCR strips in single rotor
  • Small footprint saves valuable bench space
  • Imbalance detection feature switches off the device automatically on detecting imbalance
Suitable for use in hoods and cold rooms.
Delivery information: Supplied with rotor as standard.