VWR® Non Refrigerated Microcentrifuge
Supplier: VWR Collection
VWRA521-2799EA 1700 SGD
VWRA521-2799
VWR® Non Refrigerated Microcentrifuge
Centrifuges Microcentrifuges
Benchtop microcentrifuge to spin microtubes. Widely used for clinical, molecular and bacteriological applications, high-speed microcentrifuge is ideal for small volume samples, quick spin downs, cell and bacteria
separations, isolation of macromolecules such as nucleic acids, proteins, lipids and separation of sub-cellular organelles.
  • Maximum capacity 24×1,5/2 ml
  • Variable speed setting from 500 to 13400 min⁻¹
  • Maximum RCF: 17040×g
  • Timer setting from 1 to 999 min and infinite
  • All rotors are with positional labelling, rotor cat-code, RPM/RCF(speed), rotor
More Product Information