VWR®, Doctor Centrifuge
Supplier: VWR Collection
VWRA521-2800EA 1130 SGD
VWRA521-2800
VWR®, Doctor Centrifuge
Centrifuges Microcentrifuges
The Doctor Centrifuge is designed to provide maximum user-satisfaction. The device features a powerful brushless DC motor and the cleverly designed chamber prevents temperature increases. To ensure user safety various fail-safe features have been incorporated to ensure that the operator remains safe.
  • Compact design saves valuable bench space
  • Microprocessor controlled centrifuge with variable speed and time setting
  • Imbalance detection with automatic cut off
  • Timer setting from 1 to 30 mins and infinite mode
  • Safety metal enclosure in lid
  • Lid-lock safety
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Max. capacity 6×15 ml
Max. speed 6500 min⁻¹
Timer 1 - 30 min and infinite mode
W×D×H 260×244×205 mm
Weight 4 kg