VWR®, Hematocrit Centrifuge
Supplier: VWR Collection
VWRA521-2802EA 1580 SGD
VWRA521-2802
VWR®, Hematocrit Centrifuge
Centrifuges Microcentrifuges
This hematocrit centrifuge has quality components to ensure excellent user experience. Its compact design, combined with the brushless DC motor, results in a device which is impressively powerful compared to its footprint.
  • Microprocessor controlled centrifuge with variable speed and time setting plus last run memory function
  • Precise speed setting from 500 to 12000 min⁻¹ with lowest count of 100 min⁻¹
  • HCT rotor with screw-on lid for increased contamination protection
  • Alternate RPM and RCF toggle on display during operation
  • Large 7 segment digital display
More Product Information