VWR®, Clinical Centrifuge
Supplier: VWR Collection
VWRA521-2803EA 1410 SGD
VWRA521-2803
VWR®, Clinical Centrifuge
Centrifuges Benchtop Centrifuges
This clinical centrifuge is equipped with quality components and features for a superior user expereince. With multiple rotor options, it can accommodate a wide range of sample sizes which makes it extremely versatile. The device has also been designed with a compact footprint to occupy less space relative to its functionality.
  • Microprocessor controlled for efficient performance even under heavy loads
  • Brushless DC motor for maintenance free long life
  • Favorite mode allows user to set up to 99 user defined programs
  • Enhanced airflow design to keep the
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Max. speed 4000 min⁻¹
Max. RCF 2270 ×g
Timer 1 - 999 min and infinity mode
Display Digital
Dimensions 415×355×173 mm
Weight 7,7 kg