VWR®, Microplate Centrifuge with Swing Out Rotor
Supplier: VWR Collection
VWRA521-2811EA 1280 SGD
VWRA521-2811
VWR®, Microplate Centrifuge with Swing Out Rotor
Centrifuges Benchtop Centrifuges
This microplate centrifuge can accomodate a wide range of PCR and deepwell plates which makes it extremely versatile. Because of the limited space in laboratories, it is designed with a compact footprint to occupy less space. It also has an imbalance detection feature with auto-cut off safety, to ensure smooth functioning.
  • Microprocessor controlled for efficient performance even under heavy loads
  • Digital display with speed and time setting
  • Precise RPM setting from 500 to 2200 min-¹ with lowest count of 10 min-¹
  • Enhanced airflow design to keep the
More Product Information