VWR®, Benchtop Centrifuge, Versatile Lab Genius
Supplier: VWR Collection
VWRA521-2822EA 1880 SGD
VWRA521-2822
VWR®, Benchtop Centrifuge, Versatile Lab Genius
Centrifuges Benchtop Centrifuges
The Lab Genius centrifuge offers powerful, efficient performance with years of durability. The versatility of the device is due to a wide range of rotor options which make it extemely useful across a range of laboratory requirements.
  • Microprocessor controlled for efficient performance even under heavy loads
  • Intuitive interface allows parameters to be set easily and quickly
  • Lid lock safety feature prevents lid being opened when the rotor is running 
  • One touch toggle between RCF and RPM with just a touch of a button
  • Short spin feature for ultra-fast
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Max. speed 4500 min⁻¹
Timer 30 s - 99 min and infinite mode
Display Digital
W×D×H 475×585×325 mm
Weight 23 kg