VWR® Refrigerated High Speed Microcentrifuges
Supplier: VWR Collection
VWRA521-3310EA 5260 SGD
VWRA521-3310
VWR® Refrigerated High Speed Microcentrifuges
Centrifuges Microcentrifuges
Designed to meet all your requirements, it is a multipurpose versatile laboratory centrifuge provides unmatched performance, convenience and fool proof safety. With a wide range of features like user-programmanle interface, automatic internal diagnosis, emergency lid release. This instrument is powerful and reliable centrifuge.
  • Maximum capacity of 44×1,5/2 ml – delivers up to 15000 min⁻¹
  • Brushless DC motor for maintenance-free long life
  • Temperature setting from –20 to +40 °C
  • Large back-lit LCD display for easy setting and reading of multiple parameters
More Product Information