VWR® Short Thread Vials and Septa, Wide Opening, ND9
Supplier: VWR Collection
VWRA548-2653EA 941 SGD
VWRA548-2653 VWRA548-2654 VWRA548-2660 VWRA548-2661 VWRA548-2662 VWRA548-2663
VWR® Short Thread Vials and Septa, Wide Opening, ND9
Vials
Short screw thread ND9, wide opening, thread HPLC vial with matched PP screw caps ND9, PTFE/silicone septa and micro-insert.
  • Screw vials can be used on all common autosampler
  • Short screw, long neck to make the autosampler easier to capture
  • Wider opening for easy puncture
  • PTFE/silicone septa, has excellent chemical inertness, acid and alkali resistance
  • Screw blue open top pp caps
Short screw thread ND9 vial is designed to ensure proper centering of limited volume inserts, eliminating missed injections. Available with black or blue screw caps with 9 mmMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable