Guanidinium hydrochloride ≥98%, Technical Grade

Supplier: VWR Chemicals
Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

VWRC0118-1KGEA 520 SGD
VWRC0118-1KG VWRC0118-500G VWRC0118-5KG
Guanidinium hydrochloride ≥98%, Technical Grade
Guanidinium hydrochloride

Suited for purification of nucleic acids or polypeptides from cell sources and inhibition of both RNase and protease activity.

Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ammonium Chloride ≤0.5%
Ash ≤0.4%
Moisture (KF) ≤0.3%
Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR