VWR®, Glycylglycine (Gly-Gly)

Supplier: VWR Chemicals

VWRC0137-1KGEA 1560 SGD
VWRC0137-1KG
VWR®, Glycylglycine (Gly-Gly)
Proteins and Peptides

Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH.


  • Loss on drying: ≤0,2%
  • Solubility (10%, water): Pass
  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning


Glycylglycine (Gly-Gly) contains ≤0,02% ammonia, ≤0,0001% arsenic, ≤0,2% ash, ≤0,02% chloride, ≤0,002% heavy metals (as Pb), ≤0,001% iron and ≤0,03% sulphate.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR