Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥98%, Ultra Pure Grade

Supplier: VWR Chemicals

Synonyms: Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt

VWRC0145-250MGEA 192 SGD
VWRC0145-250MG
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥98%, Ultra Pure Grade
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)

VWR offers riboneucleotides for amplification of RNA in in vitro transcription.

Formula: C₉H₁₂N₂Na₃O₁₅P₃
Storage Temperature: Freezer
MDL Number: MFCD00044310
CAS Number: 19817-92-6
EINECS: 243-347-5

Order Now

Specification Test Results

Moisture (KF) ≤13%
Purity (Dried) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR