D-(-)-Fructose ≥99%, high purity

Supplier: VWR Chemicals

Synonyms: D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose

VWRC0226-1KGEA 122 SGD
VWRC0226-1KG VWRC0226-5KG VWRC0226-12KG
D-(-)-Fructose ≥99%, high purity
D(-)-Fructose
Formula: C₆H₁₂O₆
Boiling Pt: 552 °C (1013 hPa)
Melting Pt: 100…110 °C
Density: 1,59 g/cm³ (20 °C. OECD 109)
Storage Temperature: Ambient
MDL Number: MFCD00148910
CAS Number: 57-48-7
EINECS: 200-333-3
Merck Index: 13,04295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75b7558f89edf5a510410c383e1da6b2e89ccf17&vid=3ef183bf6d7a20360c09d7ffd529fd2a227d04c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Arsenic <= 0.0001%
Calcium & Magnesium <= 0.005%
Chloride <= 0.018%
Heavy Metals (as Pb) <= 0.0005%
Loss on Drying <= 0.5%
Purity >= 99%
Residue on Ignition <= 0.5%
Sulphate <= 0.025%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR