tetra-Chloroauric(III) acid trihydrate ACS

Supplier: VWR Chemicals
Danger

Synonyms: Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate

VWRC0240-5GEA 2710 SGD
VWRC0240-5G
tetra-Chloroauric(III) acid trihydrate ACS
tetra-Chloroauric (III) acid trihydrate

Insoluble in Ether 0.1%. Purity (Au) 49.0%


Certifications: ACS Grade: Materials conforming with the specifications and procedures outlined in American Chemical Society Specifications, 9th Edition, 2000.

Formula: HAuCl₄·3H₂O
Melting Pt: 30 °C
Density: 3,9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149904
CAS Number: 16961-25-4
EINECS: 240-948-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Alkalies and Other Metals <0.2%
Insoluble in Ether ≤0.1%
Purity (Au) ≥49%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR