Sodium chloride ≥99.9% for biotechnology

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

VWRC0241-1KGEA 90 SGD
VWRC0241-1KG VWRC0241-5KG VWRC0241-500G VWRC0241-10KG VWRC0241-12KG VWRC0241-2.5KG VWRC0241-50KG
Sodium chloride ≥99.9% for biotechnology
Sodium chloride

Common source of salts for media propagation of many organisms.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Calcium ≤0.005%
DNase none detected
Heavy Metals ≤0.0005%
Iodide ≤0.002%
Iron ≤0.0002%
Loss on Drying ≤0.5%
Magnesium ≤0.005%
Nitrogen Compounds ≤0.001%
pH (5%, Water) @25 °C 5 – 9
Phosphate ≤0.0005%
Potassium ≤0.005%
Protease none detected
Purity ≥99.9%
RNase none detected
Sulphate ≤0.0004%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR