Manganese(II) sulphate monohydrate, Reagent Grade

Supplier: VWR Chemicals
Warning

VWRC0248-500GEA 250 SGD
VWRC0248-500G
Manganese(II) sulphate monohydrate, Reagent Grade
Manganese (II) sulphate monohydrate
Formula: MnSO₄·1H₂O
Melting Pt: 700 °C
Density: 2,95 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9,III
Merck Index: 13,05763

Order Now

Specification Test Results

Heavy Metals (as Pb) <0.05%
Insolubles <0.08%
Purity >98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR