Potassium permanganate ≥99% ACS

Supplier: VWR Chemicals
Danger

Synonyms: Permanganic acid potassium salt

VWRC0277-2.5KGEA 693 SGD
VWRC0277-2.5KG
Potassium permanganate ≥99% ACS
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: > 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Chloride & Chlorate <0.005%
Insolubles ≤0.2%
Purity ≥99%
Sulphate ≤0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR