Ammonium carbonate, Ultrapure

Supplier: VWR Chemicals
Warning

Synonyms: Salt of hartshorn

VWRC0305-1KGEA 187 SGD
VWRC0305-1KG
Ammonium carbonate, Ultrapure
Ammonium carbonate
Formula: (NH₄)₂CO₃
Melting Pt: 58 °C
Density: 1,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Specification Test Results

Assay (NH3) >30%
Chloride <0.0005%
Heavy Metals (as Pb) <0.0005%
Insolubles <0.005%
Iron <0.0005%
Non-volatile Matter <0.01%
Sulphur Compounds <0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR