Bromophenol blue sodium salt ACS

Supplier: VWR Chemicals

Synonyms: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt

VWRC0312-100GEA 431 SGD
VWRC0312-100G
Bromophenol blue sodium salt ACS
Bromophenol blue sodium salt
Formula: C₁₉H₉Br₄NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00013793
CAS Number: 62625-28-9
EINECS: 252-170-2

Order Now

Specification Test Results

Clarity of Solution Pass
Visual Transition Interval (pH 3.0 – 4.6) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR