Sodium pyruvate ≥99%, Reagent Grade

Supplier: VWR Chemicals

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

VWRC0342-1KGEA 294 SGD
VWRC0342-1KG VWRC0342-100G VWRC0342-250G
Sodium pyruvate ≥99%, Reagent Grade
Sodium pyruvate
Formula: CH₃COCOONa
Melting Pt: 220…230 °C
Density: 1,718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now

Specification Test Results

Free Acid ≤1
Heavy Metals (as Pb) ≤0.01%
Water Content <1%
Purity ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR