Guanidinium thiocyanate ≥99.0% for biotechnology

Supplier: VWR Chemicals
Danger

Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

VWRC0380-250GEA 178 SGD
VWRC0380-250G VWRC0380-500G VWRC0380-10KG
Guanidinium thiocyanate ≥99.0% for biotechnology
Guanidinium thiocyanate

A powerful protein denaturant used to inactivate endogenous RNases during the isolation of RNA from tissues and bacterial cultures.

Formula: CH₅N₃·CHNS
MW: 118.16 g/mol
Melting Pt: 118 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1
ADR: 8 III

Order Now

Specification Test Results

Abs.@280nm (70%, Water) ≤0.8
Abs.@300nm (6M, Water) <0.1
Abs.@410nm (6M, Water) <0.1
Copper <0.0001%
DNase none detected
Solubility (70%, Water 50° C) pass
Lead <0.0001%
Melting Point 118 – 121 °C
Protease none detected
Purity >99%
RNase none detected

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR