Lithium chloride ≥99% for biotechnology

Supplier: VWR Chemicals
Warning

VWRC0416-100GEA 214 SGD
VWRC0416-100G VWRC0416-500G
Lithium chloride ≥99% for biotechnology
Lithium chloride
Formula: LiCl
Boiling Pt: 1382 °C (1013 hPa)
Melting Pt: 614 °C
Density: 2,07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

Insolubles ≤0.01%
Iron ≤10 ppm
Purity >99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR