Lithium chloride 99 Biotechnology

Supplier: VWR Chemicals
Warning

VWRC0416-100GEA 214 SGD
VWRC0416-100G VWRC0416-500G
Lithium chloride 99 Biotechnology
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Specification Test Results

Insolubles ≤0.01%
Iron ≤10 ppm
Purity >99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR