Cholesterol, Ultrapure

Supplier: VWR Chemicals

Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol

VWRC0433-250GEA 366 SGD
VWRC0433-250G
Cholesterol, Ultrapure
Cholesterol
Formula: C₂₇H₄₆O
MW: 386.66 g/mol
Boiling Pt: 360 °C
Melting Pt: 147 °C…149 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003646
CAS Number: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221

Order Now

Specification Test Results

Loss on drying ≤0.3%
Melting point 146−150 °C
Residue on ignition ≤0.1%
Specific rotation −38 to −34°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR